Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Digital thermometer, wireless

Electronic Standard Thermometers, wireless, −20...+60 °C, ±1 °C, 0,1 °C, 60×90×30 mm
  AMARE907600
 :   Amarell
Digital thermometer, wireless
  AMARE907600
 :  Amarell
Digital thermometer, wireless
Digital thermometer, wireless

Electronic Standard Thermometers, wireless, −20...+60 °C, ±1 °C, 0,1 °C, 60×90×30 mm

 
53,30 €
   
1 * 1 ST
 

Wireless multi-channel thermometer as desktop model. Three channels connectable on one base terminal.

  • Switchable from °C to F
  • Alert option

 : Includes a remote sensor.

 
  • Range:
    −20...+60 °C
  • Resolution:
    0,1 °C
  • Description:
    Electronic Standard Thermometers, wireless
  • W×D×H:
    60×90×30 mm
  • Accuracy:
    ±1 °C
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.