ContactUsLinkComponent
Surface xleaner Titan Sanitizer
Surface xleaner Titan Sanitizer
Catalog # DIVY100884197
Supplier:  Diversey
Surface xleaner Titan Sanitizer
Catalog # DIVY100884197
Supplier:  Diversey
Specifications
  • Description:
    Titan Sanitiser, for cleaning and disinfection
  • Packed:
    Case of 12× 500 g shaker bottles
  • Pk:
    12 Bottle
Specifications
About this item
Titan Sanitizer is a chlorine based concentrated detergent sanitiser which cleans and disinfects in one step. It is effective for sanitising equipment, hard surfaces, walls, floors and instruments and can be used neat or in solution depending on the task. It kills spores at five per cent dilution.
Caution: Use biocides safely. Always read the label and product information before use.