ContactUsLinkComponent
Floor maintainer, Carefree Stride 1000
Floor maintainer, Carefree Stride 1000
Catalog # DIVY454500
Supplier:  Diversey
Floor maintainer, Carefree Stride 1000
Catalog # DIVY454500
Supplier:  Diversey
Specifications
  • Description:
    Carefree Stride 1000, floor maintainer, neutral detergent
  • Packed:
    2 × 5 l
  • Pk:
    2 Can
Specifications
About this item
Carefree Stride is a neutral, low foam detergent which is suitable for all polished (protected) and unprotected floors. It can be used in either scrubber drier or manual cleaning methods.
  • Excellent emulsification of soil, fragrance-free
  • Fast, effective removal of everyday dirt and black heel marks