Surface disinfectant, Meliseptol® rapid
Supplier: B. Braun
Certificates
About this item
Ready to use, alcoholic, surface disinfectant based on n-propanol and quaternary ammonium compound. It is active against bacteria, mycobacteria (M. terrae), yeasts and viruses (HBV, HIV, HCV, Adeno, Noro, Polio, Rota and Vaccinia).
- Activity undiluted: 1 minute
- Aldehyde- and alkylamine-free
- Fully virucidal in 10 minutes
Ordering information:
1L spray bottle (BRAU18565): Sprayhead (148-4100) must be ordered separately.