ContactUsLinkComponent
Cleaner for instruments, Helizyme
Cleaner for instruments, Helizyme
Catalog # BRAU18557
Supplier:  B. Braun
undefined
Cleaner for instruments, Helizyme
Catalog # BRAU18557
Supplier:  B. Braun
Specifications
  • Description:
    Helizyme, enzymatic manual cleaner
  • Environmentally Preferable:
  • Pk:
    1 L
Specifications
About this item
Helizyme is a pH neutral enzymatic cleaner with the combination of enzymes and surfactants, used for manual cleaning of resuable, invasive and non-invasive medical devices (surgical instruments, flexible and rigid endoscopes).
  • Excellent material compatibility
  • Suitable for ultrasonic cleaning
  • Clinically proven biofilm removal