ContactUsLinkComponent
Protective hand creams, Ligana® A Oleo-Tec
Catalog # FAWE12983001
Supplier:  FAWECO HAUTSCHUTZ
undefined
Protective hand creams, Ligana® A Oleo-Tec
Catalog # FAWE12983001
Supplier:  FAWECO HAUTSCHUTZ
Specifications
  • Description:
    Protective hand cream, Ligana® Oleo-Tec (cream A), tube
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Ligana® Oleo-Tec Creme A are protective creams (o/w emulsion) against oil-insoluble contaminants such as oils, organic solvents, non-water soluble cooling lubricants, cutting oils, and other oily substances.
  • Absorbs quickly
  • Reduces skin irritations
  • Contains glycerine and beeswax
  • Silicone-free, perfumed
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904