Slip-resistant fine resilient tapes and treads, 200 series, Safety-Walk™
Supplier: 3M
Certificates
About this item
High friction, fine resilient textured, slip-resistant material.
- For wet, barefoot-traffic areas
- Non-mineral, fine-textured, slip-resistant vinyl surface
- High performance, water-resistant, acrylate adhesive is ideal for wet conditions
- Custom shapes and sizes are available
Fine-textured, slip-resistant surface with a pressure-sensitive adhesive backing provides enhanced safety. Reverse side coated with pressure sensitive adhesive protected by a removable liner. Designed for use in wet, barefoot traffic areas. Applications include use on showers, tubs, surfboards, and around hot tubs and pools.
Frequently Bought Together