Multi-layer adhesive floor mats, Ergomat® Sticky Mats
Catalog # 115-2469
Supplier: ERGOMAT
Specifications
- Colour:White
- L×W:460×1140 mm
- Thickness:1,19 mm
- Pk:10
Specifications
About this item
Ergomat Sticky Mats provide a fast, convenient, reliable way to clean shoe soles, or other items, before entering a clean and sanitary area. They significantly reduce traffic-borne contaminants, and have proven themselves effective in a wide variety of industries, such as microelectronics, aerospace, pharmaceuticals and food processing. These disposable sticky mats consist of a stack of tear-off polyethylene films (full adhesive sheets) that effectively capture dirt and dust.
- 30 layers per mat
- Easily adhere to a frame with double-faced adhesive tape
- Uses a water-based acrylic adhesive, which has a pH of 7 to 8
- A clear release sheet protects the top sheet until it is ready to use
- Corner tabs are numbered to show the number of remaining sheets
Colour: White
Certified by RoHS to be free from heavy metals and hazardous chemicals
Certifications: Meets DIN/ASTM requirements.
Ordering information: Frame needs to be ordered separately.