Order Entry
Austria
ContactUsLinkComponent
Anti-microbial benchtop mats, Silver Bioguard
Anti-microbial benchtop mats, Silver Bioguard
Catalog # DIVESLVR-3300
Anti-microbial benchtop mats, Silver Bioguard
Catalog # DIVESLVR-3300

Specifications

  • L×W:
    559×762 mm
  • Seq:
    02
  • Thickness:
    1 mm
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

These anti-microbial mats contain silver nanoparticles to create contamination free work surface.

  • Nanoscale Silver particles bind with proteins to neutralise 99,9% of common microbes
  • Ensure consistent sample reproduction and results by significantly minimising workspace contamination
  • Silver is used extensively in medical applications where microbial contamination is a concern
  • Backed with anti-skid coating, perfect for the benchtop or culture hood