ContactUsLinkComponent
Disinfectants for instruments, Medistel
Disinfectants for instruments, Medistel
Catalog # TRSTMD307
Supplier:  TRISTEL SOLUTIONS
undefined
Disinfectants for instruments, Medistel
Catalog # TRSTMD307
Supplier:  TRISTEL SOLUTIONS
Specifications
  • Description:
    Medistel, instrument disinfectant
  • Seq:
    01
  • Packed:
    2 polyboxes of 20×50 ml sachets in case
  • Pk:
    40
Specifications
About this item
Medistel is used for chemical reprocessing of thermo-sensitive surgical instruments, flexible or rigid endoscopes and devices. It is suitable for both human and veterinary applications for disinfection of surgical instruments and medical devices. It is designed for open tank use and also suitable for other medical apparatus including ultrasonic baths.
  • Disinfects in 10 minutes and sterilises in 30 minutes
  • Dilution for use: 1:20
  • Compliant with BS EN ISO 9001:2000 and BS EN ISO 13485:2003
  • Conforms to medical directive 93/42/EEC requirements