Order Entry
Austria
ContactUsLinkComponent
Dispenser bottle, LDPE
Dispenser bottle, LDPE
Catalog # HUEN844200
Supplier:  hünersdorff GmbH
undefined
Dispenser bottle, LDPE
Catalog # HUEN844200
Supplier:  hünersdorff GmbH

Specifications

  • Capacity:
    50 ml
  • Height:
    79 mm
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

LDPE, natural, food-safe.

  • Resistant to acids and chemicals
  • Dosing by pressure on the bottle
  • With closed nozzle and protective cap

For glue, oil, and more, oval shape.