ContactUsLinkComponent
Wipes, pre-saturated
Wipes, pre-saturated
Catalog # EPTA6-LS964-685
undefined
Wipes, pre-saturated
Catalog # EPTA6-LS964-685
Specifications
  • Packed:
    100/canister
  • L×W:
    152×216 mm
  • Pk:
    1.200
Specifications
About this item
These polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
  • Perforated roll pre-wetted with alcohol
  • Canister with spring loaded lid prevents dying out and facilities dispensing
  • Alcohol saturates and evaporates quickly
  • Made in the USA
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904