ESD safety shoes, lace-up, A 100 S981
Catalog # ATLAESDA100/42
Supplier: ATLAS - the shoe company
Specifications
- Shoe size:42
- Colour:Amber
- Pk:1 PAIR
Specifications
About this item
Sneaker style ESD safety shoes made from full-grained calf leather and Sneaker Line sole (3,0 mm profile, oil, acid and petrol resistant, anti-static).
- Skin friendly, abrasion resistant aktiv-X lining for enhanced breathability
- Removable insoles; orthopedic insoles possible
- 3D cushioning system: pressure relieving insole, cushioning comfort midsole, dynamic outsole system made from MPU
- Protective steel toecap (200 J)
- Electrical contact resistance of 1×10⁵ to 3,5×10⁷ Ohm
Colour: Brown
Certifications: EN 61340 (ESD); DGUV 112-191; EN ISO 20345; S2