ContactUsLinkComponent
Scroll pumps, XDS-C series
Scroll pumps, XDS-C series
Certificates
Scroll pumps, XDS-C series
Specifications
  • Catalog No:
  • 181-0365
  • 181-0363
  • Model:
  • XDS10-C
  • XDS5-C
  • Final pressure with gas ballast:
  • 0,5 mbar
  • 0,5 mbar
  • Connection on pressure side (EX):
  • NW25
  • NW25
  • Connection on suction side (IN):
  • NW25
  • NW25
  • Power connection:
  • 230 V, 50/60 Hz
  • 230 V, 50/60 Hz
  • Motor output:
  • 0,3 kW
  • 0,3 kW
  • Weight:
  • 24,5 kg
  • 23 kg