ContactUsLinkComponent
Hand disinfectants, Manusept® basic
Hand disinfectants, Manusept® basic
Catalog # BODL975612
Supplier:  Bode
undefined
Hand disinfectants, Manusept® basic
Catalog # BODL975612
Supplier:  Bode
Specifications
  • Description:
    Hand disinfectant, Manusept® basic, 500 ml bottle
  • Seq:
    04
  • Pk:
    1
Specifications
About this item
The disinfectant is useful for hygienic and surgical disinfection of the hands for normal and sensitive skin.
  • Active against bacteria, fungi, and enveloped viruses
  • Good skin tolerability
  • Ethanol based
  • Colourant- and fragrance-free
Caution: Use biocides safely. Always read the label and product information before use.
* These articles are only available in Germany.
*The product(s) on this page are outside the scope of the EU Directive 2019/904