Roll mop, PolyTack®
Supplier: CONNECTICUT CLEANROOM
Jump to:
About this item
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
- Easy to use hand or pole attachments
- Plastic inner core insures cleanliness
- Anti-microbial agent providing protection against bacterial growth
4 Options Available Below
Product Details & Documents
Tacky roller, with foam or film for cleaning walls, floors, ceiling and hard to reach areas.
- Easy to use hand or pole attachments
- Plastic inner core insures cleanliness
- Anti-microbial agent providing protection against bacterial growth
Product Family Options
Product Information
- PackedAvailabilityPrice
- CONNPR-18Tacky roller, film for flat surfaces, 457,2 mmTacky roller, film for flat surfaces, 457,2 mm40 sheets/rollLog in for availability1 * 1 Rolle82,70 €
- CONNPR-9FTacky roller, foam for contoured surfaces, 228,6 mmTacky roller, foam for contoured surfaces, 228,6 mm40 sheets/rollLog in for availability1 * 1 Rolle56,60 €
- CONNPR-18FTacky roller, foam for contoured surfaces, 457,2 mmTacky roller, foam for contoured surfaces, 457,2 mm20 sheets/rollLog in for availability1 * 1 Rolle82,70 €
Mop handle
Product Information