Sleeve protector, PP/PE, Sekuroka®
ROTH8202.2
- Garment application:Laboratory
- Colour:White
- Length:580 mm
- Pk:10 PAIR
These protective sleeves with elastic bands are made of PP with PE coating.
- Anti-static, lint-free
- Tearproof and water repellent