Order Entry
Austria
ContactUsLinkComponent
Sample vials for coulter counter
Sample vials for coulter counter
Catalog # SARS73.650.500
Supplier:  SARSTEDT
Sample vials for coulter counter
Catalog # SARS73.650.500
Supplier:  SARSTEDT

Specifications

  • Cap included:
    Yes
  • Ø×H:
    34×60 mm
  • Capacity:
    20,00 ml
  • Seq:
    01
  • Pk:
    500

Specifications

About this item

Sample vials for Coulter Counter with enclosed caps. Vials are made from PS, caps from LDPE.

  • Flat base
Packaging: 250 vials and caps are packed in the bag.