Orders LinkContactUsLinkComponent
Cryoboxes
Supplier:   RATIOLAB
Cryoboxes
Supplier:  RATIOLAB
Certificates
9 Options Available Below
Product Details & Documents
PC, coloured, with clear lid.
  • Temperature-resistant from −190 to 121 °C
  • Drain bores at the bottom for dew liquids
  • By stacking corners the boxes are suitable for safe transport
With slip lid and variable grid insert.
Product Family Options
Product Information
  • Compartments
    For
    Colour
    W×D×H ext.
    Availability
    Price
  • 9×9
    Cryotubes 1,2 - 2 ml
    Green
    132×132×52 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,14 €
    55,14 €
    1 * 5 ST
       55,14 €
  • 9×9
    Cryotubes 1,2 - 2 ml
    Red
    132×132×52 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,14 €
    55,14 €
    1 * 5 ST
       55,14 €
  • 10×10
    Cryotubes 1,2 - 2 ml
    Blue
    132×132×52 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,78 €
    55,78 €
    1 * 5 ST
       55,78 €
  • 10×10
    Cryotubes 1,2 - 2 ml
    Green
    132×132×52 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,78 €
    55,78 €
    1 * 5 ST
       55,78 €
  • 10×10
    Cryotubes 1,2 - 2 ml
    Red
    132×132×52 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,78 €
    55,78 €
    1 * 5 ST
       55,78 €
  • 10×10
    Cryotubes 1,2 - 2 ml
    Yellow
    132×132×52 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 55,78 €
    55,78 €
    1 * 5 ST
       55,78 €
  • 9×9
    Cryotubes 3 - 5 ml
    Blue
    132×132×94 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 58,69 €
    58,69 €
    1 * 4 ST
       58,69 €
  • 9×9
    Cryotubes 3 - 5 ml
    Green
    132×132×94 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 58,69 €
    58,69 €
    1 * 4 ST
       58,69 €
  • 9×9
    Cryotubes 3 - 5 ml
    Red
    132×132×94 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 58,69 €
    58,69 €
    1 * 4 ST
       58,69 €