ContactUsLinkComponent
Clamps, three pronged
Clamps, three pronged
Catalog # BOCH5565
Supplier:  BOCHEM
undefined
Clamps, three pronged
Catalog # BOCH5565
Supplier:  BOCHEM
Specifications
  • Spanning width:
    0 - 100 mm
  • Description:
    Clamps, three pronged
  • Length:
    140 mm
  • Material:
    Aluminium
  • Pk:
    1
Specifications
About this item
Three pronged retort clamp made of aluminium.
  • Fingers of the clamps are covered with a layer of cork to protect clamped items