Lens tissue papers, MN 13
Lieferant: MACHEREY-NAGEL
Zertifikate
Über diesen Artikel
Very thin and soft filtration paper for cleaning optical glasses and cuvettes.
Frequently Bought Together
Very thin and soft filtration paper for cleaning optical glasses and cuvettes.