ContactUsLinkComponent
Single wafer shipper, for 300 mm wafer handling, H 93 series
Single wafer shipper, for 300 mm wafer handling, H 93 series
Catalog # 216-1588
Supplier:  ENTEGRIS
undefined
Single wafer shipper, for 300 mm wafer handling, H 93 series
Catalog # 216-1588
Supplier:  ENTEGRIS
Specifications
  • Description:
    Shipper, PC, clear
  • Seq:
    04
  • Pk:
    5
Specifications
About this item
The H 93 single wafer shipper is designed to securely retain the wafer and minimise particle generation. Stackable with multiple closures, each shipper holds one 300 mm wafer facedown contacting only the wafer’s edges.
  • Secure, replaceable cushions for minimised particle generation
  • Single latch for secure closing and easy opening
  • Stackable for storage efficiency
  • Clear PC construction for visual inspection without opening
Provides secure wafer protection in a convenient, easy to use package.
Please contact Avantor for complete range of products and options.
Caution: Articles may not be available in all countries. Please contact your local Avantor office.