Pasteur pipettes, disposable, LDPE
Supplier: SEMADENI
Jump to:
About this item
LDPE, disposable, non-sterile.
- Almost unbreakable transfer pipettes
- Good chemical resistance and can be used at low temperatures
- Pipettes are made from one piece: No risk of leakage
- Dustproof packaged in convenient standard boxes
4 Options Available Below
Product Details & Documents
LDPE, disposable, non-sterile.
- Almost unbreakable transfer pipettes
- Good chemical resistance and can be used at low temperatures
- Pipettes are made from one piece: No risk of leakage
- Dustproof packaged in convenient standard boxes