Analytical HPLC columns, XTerra® RP18
Lieferant: WATERS
Zertifikate
Über diesen Artikel
XTerra® RP18 columns are general purpose, hybrid-based, reversed phase C18 columns that incorporate Waters patented embedded polar group technology to improve peak shape performance for highly basic compounds. These first generation columns provide superior pH stability compared to a silica-based column.
Frequently Bought Together