ContactUsLinkComponent
MEM vitamins 100X concentrate
MEM vitamins 100X concentrate
Catalog # X0556-100
Supplier:  Biowest
CAS Number:  
undefined
MEM vitamins 100X concentrate
Catalog # X0556-100
Supplier:  Biowest
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    MEM vitamins, 100X, without L-glutamine (liquid)
Specifications
About this item
Concentrated solutions of amino acids, vitamins or non essential amino acids can be added to a basal medium to enrich it and the resulting medium used as a classical MEM or BME. The final concentration of the MEM or BME must be 1X.