Order Entry
Austria
Orders LinkContactUsLinkComponent
Microincinerators, e-Loop

Microincinerators, e-Loop

Supplier:   BioTool
Microincinerators, e-Loop
Supplier:  BioTool
Certificates
Microincinerators, e-Loop

2 Options Available Below

Product Details & Documents

Electrical sterilisation systems which are suitable for sterilising loops, needles and instruments. Ideal for use in laboratories where using gases or naked flames is not permitted. The e-Loop has a wear resistant quartz glass tube which is protected by a perforated stainless steel outer design. Choice of two units with different tube diameters, the larger is ideal for sterilising small instruments such as forceps and scissors. The heating system provides uniform heat. The opening eliminates the potential aerosol of pathogenic microorganisms.

  • Loops and needles sterilised in 5 to 7 s
  • Integral spray protector prevents hazardous gases, flames and infectious splatter harming user
  • Sterilisation tube has self-locking angular adjustment, there are seven different positions
  • Tray for three instruments
  • Residual heat warning to prevent burns

Bestellinformation: Optional accessories include a holder specifically for BioTool inoculation loop holders, a stainless steel tray which is positioned on the left hand side of the steriliser and stainless steel inoculation loop holders for loops with wire Ø 0,6 to 1 mm.

Specifications for Product Family

Specifications

  • Catalog No:
  • 710-1471
  • 710-1472
  • Temperature range:
  • 900 - 950 °C
  • 900 - 950 °C
  • Ø×L:
  • 15×118 mm
  • 21×161 mm
  • Power supply:
  • 120 W
  • 120 W
  • W×D×H:
  • 175×139×129 mm
  • 175×160×173 mm
  • Weight:
  • 1,1 kg
  • 1,6 kg
  • Model:
  • 710-1471
  • 710-1472

Specifications

Product Family Options

Product Information

  • Model
    Availability
    Price
  • 710-1471
    e-Loop with 15 mm tube diameter
    e-Loop with 15 mm tube diameter
    710-1471
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 775,00 €
    775,00 €
    1 * 1 ST
       775,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    710-1471
    Temperature range
    900 - 950 °C
    Ø×L
    15×118 mm
    Power supply
    120 W
    W×D×H
    175×139×129 mm
    Weight
    1,1 kg
    Model
    710-1471
  • 710-1472
    e-Loop Plant with 21 mm tube diameter
    e-Loop Plant with 21 mm tube diameter
    710-1472
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 836,00 €
    836,00 €
    1 * 1 ST
       836,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    710-1472
    Temperature range
    900 - 950 °C
    Ø×L
    21×161 mm
    Power supply
    120 W
    W×D×H
    175×160×173 mm
    Weight
    1,6 kg
    Model
    710-1472
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stainless steel tray for positioning on left hand side
    Stainless steel tray for positioning on left hand side
    710-1575
    Log in for availability
    1 * 1 ST - 61,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Holding device for BioTool loop holders
    Holding device for BioTool loop holders
    710-1473
    Log in for availability
    1 * 1 ST - 54,90 €

Product Information

  • Catalog #
    Availability
    Price
  • Quartz glass heating element for e-Loop 230 V
    Quartz glass heating element for e-Loop 230 V
    710-1573
    Log in for availability
    1 * 1 ST - 339,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Spare ceramic annealing tube for e-Loop standard, all voltages
    Spare ceramic annealing tube for e-Loop standard, all voltages
    710-1574
    Log in for availability
    1 * 1 ST - 89,90 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.