ContactUsLinkComponent
Microplate sealing tape
Microplate sealing tape
Catalog # 738-3002
   Low Manufacturing Impact
undefined
Microplate sealing tape
Catalog # 738-3002
   Low Manufacturing Impact
Specifications
  • Description:
    PVC sealing tape
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (Certificate Registration No. EMS 623400)
  • Pk:
    100
Specifications
About this item
Adhesive sealing tape for microplates. Temperature range: −30 to +80 °C.