ContactUsLinkComponent
Tubing coupling
Tubing coupling
Catalog # BURK8704-0608
Supplier:  BURKLE
undefined
Tubing coupling
Catalog # BURK8704-0608
Supplier:  BURKLE
Specifications
  • Tubing int.Ø:
    5-7 mm
  • Description:
    Hose connector, conical, attachable
  • Pk:
    10
Specifications
About this item
Attachable, conical hose couplings, made of PE, for pressure less operation.
  • Straight
  • Good chemical resistance
  • Good temperature resistance