Liquid chromatography sorbents, Polyamide 6
Polyamide 6 adsorbents for column chromatography.
- Polyamide 6 (ε-polycaprolactam)
- Chromatographic quality
- Separation mechanism mainly based on hydrogen bonds
Recommended application: separation of phenolic compounds (e.g., isolation of natural products), carboxylic acids and aromatic nitro compounds.
Product Family Options
- DescriptionPk
- Polyamide CC 6, with particle size of <0,07 mm1 kg1 * 1 kg459,00 €
- Polyamide CC 6, with particle size of <0,07 mm5 kg1 * 5 kg2 095,00 €
- Polyamide CC 6, with particle size of 0,05 - 0,16 mm1 kg1 * 1 kg459,00 €
- Polyamide CC 6, with particle size of 0,05 - 0,16 mm5 kg1 * 5 kg2 095,00 €
- Polyamide CC 6, with particle size of 0,10 - 0,30 mm1 kg1 * 1 kg414,00 €
- Polyamide CC 6, with particle size of 0,10 - 0,30 mm5 kg1 * 5 kg1 885,00 €