Wipe down table
Supplier: Palbam
Jump to:
About this item
The wipe down table is designed to support the wipe down procedure. Includes a hole in the table top for easy waste disposal.
The cabinet under the table and under the waste disposal hole has a removable plastic bin included inside.
In stainless steel 304 with electropolished finish.
2 Options Available Below
Product Details & Documents
The wipe down table is designed to support the wipe down procedure. Includes a hole in the table top for easy waste disposal.
The cabinet under the table and under the waste disposal hole has a removable plastic bin included inside.
In stainless steel 304 with electropolished finish.
Documents
Palbam Class - Catalog 2018
Product Literature
Product Family Options
Product Information
- W×D×HAvailabilityPrice
- 134-0489Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic binWipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin1200×600×920 mmLog in for availability1 * 1 ST
- 132-1091Wipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic binWipe down table, with waste disposal hole on top and a cabinet under the table with removable plastic bin900×600×900 mmLog in for availability1 * 1 ST4 210,00 €