Order Entry
Austria
ContactUsLinkComponent
Test tubes with screw cap, sterile
Test tubes with screw cap, sterile
Catalog # GREI164162_1000
Supplier:  Greiner Bio-One
   Low Manufacturing Impact
Test tubes with screw cap, sterile
Catalog # GREI164162_1000
Supplier:  Greiner Bio-One
   Low Manufacturing Impact

Specifications

  • Ext.Ø:
    16,8 mm
  • Capacity:
    12 ml
  • Length:
    100 mm
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    ISO 14001 (241002); ISO 50001 (241002)
  • Pk:
    1.000

Specifications

About this item

Polystyrene (PS), clear.

  • Conical bottom, skirted
  • Good thermal resistance
  • Good chemical resistance

Universal tubes for a wide range of applications.