ContactUsLinkComponent
Barcode reader, Nunc™
Barcode reader, Nunc™
Catalog # NUNC330050
undefined
Barcode reader, Nunc™
Catalog # NUNC330050
Specifications
  • Description:
    Barcode scanner, for use with any USB port
  • Interfaces:
    USB
  • Pk:
    1
Specifications
About this item
This durable barcode reader is ideal for simple track sampling. It is for use with any USB slot.
  • 'Plug and Play' function can automatically switch to read codes: 128A, 128B, 128C, 39 and interleave ²/₅ Convenient and easy to use with an Excel spread sheet
  • Scanner is a contact scanner which means it must be in contact with the barcode to read