Orders LinkContactUsLinkComponent
Pass through cabinets
Pass through cabinets
Supplier:   Palbam
Pass through cabinets
Supplier:  Palbam
Certificates
Pass through cabinets
Pass through cabinets
2 Options Available Below
Product Details & Documents
Pass Through cabinet allows products and materials to be transferred between different rooms. Reduces unnecessary walking and entering/exiting cleanroom.
  • Single or double versions available
  • Sealing gaskets and compression handles ensure firm closure
  • Tempered Glass or anti static PVC windows available
  • Mechanical or electronic interlocks available
  • Can be customised to requirements
Fabrication material: 304 stainless steel with electropolished finish.
Product Family Options
Product Information
  • Doors
    W×D×H
    Availability
    Price
  • PALBPT-604060
    Pass through cabinet, with static dissipative window, sealing gasket and compression handle
    Pass through cabinet, with static dissipative window, sealing gasket and compression handle
    2
    600×400×600 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • PALBPT-706070MS
    Pass through cabinet, with static dissipative window, sealing gasket and compression handle
    Pass through cabinet, with static dissipative window, sealing gasket and compression handle
    2
    700×600×700 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST