Desiccator cabinets
About this item
Bench top N₂ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets cut nitrogen expenses by up to 80%, automate clean, dry benchtop storage eliminating moisture & oxygen related degradation and optimise yields. An appropriate flow of nitrogen is introduced into the upper section of the desiccator; the heavier air is then purged out of the bottom. Ideal for use with semiconductor components, biological and pharmaceutical samples, and other sensitive materials. Fully integrated turnkey system takes 30 s to install.
- Built in RH% sensor and display panel
- Easy to use control panel allows user to enter the required RH% level
- Can be used as a single unit on bench top or shelf or as units of two or three in dedicated shelf units
- N₂ desiccator cabinets minimise the N₂ usage and therefore minimise N₂ leakage into the cleanroom atmosphere
Delivery information: Typically available from stock for direct shipment from the supplier.
3 Options Available Below
Product Details & Documents
Bench top N₂ purge desiccator cabinets made from stainless steel 304, units have an electropolished finish and static dissipative PVC window. Cabinets cut nitrogen expenses by up to 80%, automate clean, dry benchtop storage eliminating moisture & oxygen related degradation and optimise yields. An appropriate flow of nitrogen is introduced into the upper section of the desiccator; the heavier air is then purged out of the bottom. Ideal for use with semiconductor components, biological and pharmaceutical samples, and other sensitive materials. Fully integrated turnkey system takes 30 s to install.
- Built in RH% sensor and display panel
- Easy to use control panel allows user to enter the required RH% level
- Can be used as a single unit on bench top or shelf or as units of two or three in dedicated shelf units
- N₂ desiccator cabinets minimise the N₂ usage and therefore minimise N₂ leakage into the cleanroom atmosphere
Delivery information: Typically available from stock for direct shipment from the supplier.
Documents
Palbam Class - Catalog 2018
Product Literature
Product Family Options
Product Information
- WithW×D×H ext.AvailabilityPrice
- PALBUCD-604040Bench top N₂ purge desiccator cabinet, with static dissipative PVC windowBench top N₂ purge desiccator cabinet, with static dissipative PVC windowRH% sensor and display panel600×400×400 mmLog in for availability1 * 1 ST5 510,00 €
- PALBUCD-604060Bench top N₂ purge desiccator cabinet, with static dissipative PVC windowBench top N₂ purge desiccator cabinet, with static dissipative PVC windowRH% sensor and display panel600×400×600 mmLog in for availability1 * 1 ST
- PALBUCD805050Bench top N₂ purge desiccator cabinet, with static dissipative PVC windowBench top N₂ purge desiccator cabinet, with static dissipative PVC windowRH% sensor and display panel800×500×500 mmLog in for availability1 * 1 ST
Accessories
Product Information
- Catalog #ForAvailabilityPrice
- Shelf, solid stainless steel, 800×500 mmShelf, solid stainless steel, 800×500 mmPALBDS8050Desiccator UCD8050Log in for availability1 * 1 ST
Product Information
- Catalog #ForAvailabilityPrice
- Shelf, solid stainless steel, 600×400 mmShelf, solid stainless steel, 600×400 mmPALBDS-6040Desiccator UCD6040Log in for availability1 * 1 ST
Product Information
- Catalog #ForAvailabilityPrice
- Desiccator stand, stainless steel, for three unitsDesiccator stand, stainless steel, for three unitsPALBDSTAND604040-3Desiccator UCD604040Log in for availability1 * 1 ST