Orders LinkContactUsLinkComponent
Sample cups for analyser
Sample cups for analyser
For Coulter Counter analysers, with cap, 20 ml, Bag of 250 or 1250/case
Catalog # 391-3720
Supplier:   Greiner Bio-One
Sample cups for analyser
Catalog # 391-3720
Supplier:  Greiner Bio-One
Sample cups for analyser
For Coulter Counter analysers, with cap, 20 ml, Bag of 250 or 1250/case
Price
176,00 €
Log in for availability
1 * 1.250 ST
Product Details & Documents
Cups made of PS.
  • Conical bottom and supported with skirts
  • Good chemical resistance
  • Good temperature resistance
Suitable for analytical systems, such as Technicon, Gemsaec, Centrifichem, Hitachi, TOA and Coulter analysers.
Specifications
  • Packed:
    Bag of 250 or 1250/case
  • Capacity:
    20 ml
  • Description:
    For Coulter Counter analysers, with cap
  • Seq:
    05