Order Entry
Austria
ContactUsLinkComponent
Bouffant cap, BioClean™ BBC
Bouffant cap, BioClean™ BBC
Catalog # 113-8790
Supplier:  Nitritex
undefined
Bouffant cap, BioClean™ BBC
Catalog # 113-8790
Supplier:  Nitritex

Specifications

  • Ø:
    50×52 cm
  • Garment application:
    Cleanroom
  • Colour:
    White
  • Pk:
    1.000

Specifications

About this item

These lightweight cleanroom bouffant caps are made of non woven PP.

  • Hypoallergenic, soft fabric
  • Elasticated double stitching
  • Latex-free
  • Compatible with a variety of environments

Density: 10 g/m²