Order Entry
Austria
ContactUsLinkComponent
Disposable particle filtering respirators, FFP2, P201, BizTex®
Disposable particle filtering respirators, FFP2, P201, BizTex®
Catalog # PRTWP201WHR
Supplier:  Portwest
undefined
Disposable particle filtering respirators, FFP2, P201, BizTex®
Catalog # PRTWP201WHR
Supplier:  Portwest

Specifications

  • Type:
    P201
  • Pk:
    10
  • With valve:
    +
  • Class:
    FFP2

Specifications

About this item

BizTex consists of a polypropylene substrate to which a laminated microporous PE film is applied (PP/PE). The outer layer of fabric prevents the passage of liquid molecules inwards yet permits the vapour molecules that build up inside to pass through. Mask protects against fine toxic dust, fumes and water based mists.

  • Adjustable nose bridge
  • Sweat absorbing nose foam (silicone/latex-free)
  • Twin elastic headband
  • Colour coded to indicate performance level
  • With easy flow exhalation valve
  • Can be used in concentrations of contaminants up to 10 times WEL
Certifications: EN 149:2001+A1:2009