Orders LinkContactUsLinkComponent
Safety cutters, retractable, KN40
Safety cutters, retractable, KN40
Safety cutter, KN40
Catalog # PRTWKN40BLU
Supplier:   Portwest
Safety cutters, retractable, KN40
Catalog # PRTWKN40BLU
Supplier:  Portwest
Safety cutters, retractable, KN40
Safety cutters, retractable, KN40
Safety cutter, KN40
Price
17,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Carbon steel blade, handle made of zinc alloy.
This cutter features a blade that retracts when it loses contact with material being cut. Primarily for use in warhousing, distribution or constructtion environments. Pack contains 10 replacement blades.
Certifications: TÜV + GS approved
Specifications
  • Description:
    Safety cutter, KN40
Accessories
Product Information
  • Catalog #
    Material
    Availability
    Price
  • Replacement blades for KN40, two cutting edges
    Replacement blades for KN40, two cutting edges
    PRTWKN91NCR
    Carbon steel
    Log in for availability
    1 * 500 ST - 104,00 €