Handheld refractometers
0 - 80 °Brix, 0,5 °Brix
Catalog # 635-0081
Supplier: EUROMEX
Jump to:
Product Details & Documents
Single dual or triple scales, analogue, handheld refractometers.
- Automated temperature compensation (ATC)
- Calibrated at 20 °C except for RF.5190
- Requires only a few drops of liquid
Specifications
- Messbereich:0 - 80 °Brix
- Seq:07
- Präzision:0,5 °Brix