Order Entry
Austria
ContactUsLinkComponent
Microcentrifuge tubes
Microcentrifuge tubes
  BOET02.013.0100
undefined
Microcentrifuge tubes
  BOET02.013.0100

 

  • Packed:
    1000/bag
  • Type:
    without lid
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Natural
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    1.000

 

 

Made from selected high-quality and biologically inert polypropylene.

  • High resistance to temperatures and chemicals
  • Autoclavable at 121°C/2,1 bar (cap not closed)
  • Place for markings on label area and flat cover
  • Perfect closure of leak free cap, due to the design of the lid
  • Membrane can be punctured by sampling-needles
  • Graduation
  • Round inside bottom for easy recovery of pellets or fluids

Without caps.