Order Entry
Austria
ContactUsLinkComponent
Transport and Storage Container
Transport and Storage Container
Catalog # SEMA1940
Supplier:  SEMADENI
undefined
Transport and Storage Container
Catalog # SEMA1940
Supplier:  SEMADENI

Specifications

  • W×D×H ext.:
    600×800×500 mm
  • Capacity:
    180 l
  • Description:
    With four swivel castors of Ø 75 mm
  • Pk:
    1

Specifications

About this item

Transport and storage containers, made of PP, grey, with reinforced rim and two handles. They are not suitable for liquids.