Order Entry
Austria
ContactUsLinkComponent
Syringes with Luer fittings, PP, medi syst
Syringes with Luer fittings, PP, medi syst
Artikelnr. SEMA0900
Supplier:  SEMADENI
undefined
Syringes with Luer fittings, PP, medi syst
Artikelnr. SEMA0900
Supplier:  SEMADENI

Specifications

  • Capacity:
    200 ml
  • Description:
    Syringe, large, non sterile, tapered tip, tip Ø 6 - 12 mm, tip length 40 mm
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

Syringes with three finger rings for a comfortable, precise and residue-free dosage.

  • Graduated scale engraved
  • Easy to disassemble and clean
  • Perfect sealing and easy piston stroke
  • Equipped with a Luer fitting and conical tip
  • With lid lock preventing the piston from jumping

Low pressure is sufficient to inject or dose products, even viscous.