Orders LinkContactUsLinkComponent
Pipette tips, universal
Pipette tips, universal
Supplier:   BOETTGER PAUL
Pipette tips, universal
Supplier:  BOETTGER PAUL
Certificates
Pipette tips, universal
Pipette tips, universal
3 Options Available Below
Product Details & Documents
These tips are made from selected, hydrophobic PP.
  • Optimal surface quality
  • Precise and centric opening of tip
  • For optimal use on single and multi-channel pipettes
  • Autoclavable at 121 °C/2,1 bar
  • High accuracy
Product Family Options
Product Information
  • Tip style
    Volume
    Length
    Tip colour
    Packed
    Availability
    Price
  • Extended
    0,1 - 20 µl
    40,5 mm
    Transparent
    Bulk (1000/bag)
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST - 155,00 €
    155,00 €
    1 * 10.000 ST
       155,00 €
  • Standard
    2 - 200 µl
    52,6 mm
    Yellow
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 13,70 €
    13,70 €
    1 * 1.000 ST
       13,70 €
  • Standard
    50 - 1000 µl
    70,5 mm
    Blue
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 17,10 €
    17,10 €
    1 * 1.000 ST
       17,10 €