ContactUsLinkComponent
Cleanroom wipes, pre-saturated, AlphaSat® with Vectra® AlphaSorb® 10
Cleanroom wipes, pre-saturated, AlphaSat® with Vectra® AlphaSorb® 10
Catalog # TEXWTX1057
Supplier:  Texwipe
Cleanroom wipes, pre-saturated, AlphaSat® with Vectra® AlphaSorb® 10
Catalog # TEXWTX1057
Supplier:  Texwipe
Specifications
  • Packed:
    40 wipers/reclosable bag, 12 bags/case
  • Description:
    TX1057
  • L×W:
    230×230 mm
  • Pk:
    480
Specifications
About this item
AlphaSat® with Vectra® AlphaSorb®10 incorporates ITW Texwipe's ultraclean AlphaSorb 10 sealed-border wipes (100% polyester) with 10% semiconductor-grade isopropyl alcohol (0,2 µm filtered) in an easy-to-use flatpack package.
  • Superior strength and abrasion resistance in a continuous-filament knitted substrate
  • Low particle generation and low extractable levels help prevent product and cleanroom contamination
  • Eliminates problems associated with purchasing, storing, filtering and diluting alcohol
  • Reduces alcohol consumption and eliminates over-spray of alcohol
Ideal for cleaning production equipment and environmental surfaces for reduction of particulate contamination, for wipedown of articles to be passed or carried into cleanrooms, for process chamber wipedowns in semiconductor processing and for cleaning fingerprints and light soils from sensitive medical devices prior to packaging and terminal sterilisation.
Packaging: Packaged in a cleanroom environment. Wipes are protected in a sealed outer bag for contamination-free introduction into the cleanroom.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904.