Orders LinkContactUsLinkComponent
Temperature data loggers
Temperature data loggers
Supplier:   COMARK
Temperature data loggers
Supplier:  COMARK
Certificates
4 Options Available Below
Product Details & Documents
These data loggers measure both humidity and temperature when monitoring food in storage, transit or processing as well as pharmaceutical and general industrial applications.
  • Large memory capacity and single button control
  • Ability to log over multiple periods
  • Windows™ based evolution software for fast data download and analysis
  • LED indication of active logging and temperature or humidity alarm conditions
  • LCD displays on selected models for instant checks on current readings and alarms
  • Additional beeper warning of temperature and humidity alarm conditions
Models with thermistor sensors offer highest accuracy. K or T-Type thermocouples provide a wide measurement range. With the addition of the N2000BOX, the N2012 can measure up to five separate channels with a single logger using four external probes and one internal sensor. With the N2000ADP/K, the N2014 can measure up to four channels using three external thermocouple probes and one internal sensor.
Certifications: Tough, moulded cases, dust and waterproof to IP67 standards; BS EN 60529; IEC 529 certified.
Specifications for Product Family
Specifications
  • Catalog No:
  • COMAN2011
  • COMAN2012
  • COMAN2013
  • COMAN2014
  • Sensor:
  • Thermistor
  • Thermistor
  • T: Thermistor
    RH: Capacitive
  • Int.: Thermistor
    Ext.: Types T/K thermocouple
  • Display:
  • LCD
  • LCD
  • LCD
  • LCD
  • Battery life:
  • 5 years
  • 5 years
  • 5 years
  • 5 years
  • Recording intervals:
  • 1 s - 99 hr
  • 1 s - 99 hr
  • 1 s - 99 hr
  • 2 s - 99 hr
  • Response time:
  • T90, 15 - 30 min
  • T90, 15 - 30 min
  • T90, 15 - 30 min
  • T90, 15 - 30 min
  • W×D×H:
  • 88×80×35 mm
  • 88×80×35 mm
  • 88×80×35 mm
  • 88×80×35 mm
  • Data memory - No. of readings:
  • 16000
  • 16000
  • 16000
  • 16000
  • Batteries:
  • 1×AA size, 3,6 V lithium battery
  • 1×AA size, 3,6 V lithium battery
  • 1×AA size, 3,6 V lithium battery
  • 1×AA size, 3,6 V lithium battery
Specifications
Product Family Options
Product Information
  • Range
    Accuracy
    Resolution
    Availability
    Price
  • COMAN2011
    Temperature data logger with internal thermistor
    Temperature data logger with internal thermistor
    T: –40…+70 °C
    T: ±0,5 °C (–25...+50 °C); ±1,0 °C (–40...+70 °C)
    T: 0,1 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 491,00 €
    491,00 €
    1 * 1 ST
       491,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    COMAN2011
    Sensor
    Thermistor
    Display
    LCD
    Battery life
    5 years
    Recording intervals
    1 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
  • COMAN2012
    Temperature data logger with one internal and four external temperature sensors
    Temperature data logger with one internal and four external temperature sensors
    T: –40…+70 °C Int.;
    T: –40…+150 °C Ext.
    T: ±0,5 °C (–25…+50 °C); ±1,0 °C (–40…+70 °C); ±2,0 °C(70...150 °C)
    T: 0,1 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 633,00 €
    633,00 €
    1 * 1 ST
       633,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    COMAN2012
    Sensor
    Thermistor
    Display
    LCD
    Battery life
    5 years
    Recording intervals
    1 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
  • COMAN2013
    Single-use temperature logger, with humidity alarm
    Single-use temperature logger, with humidity alarm
    T: –20…+60 °C
    RH: 0 - 97 %
    T: ±0,5 °C (–20...+50 °C); ±0,5 °C (50...60 °C); ±1 °C otherwise
    RH: ±3% (–20...+60 °C)
    T: 0,1 °C
    RH: 0.1 %
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 897,00 €
    897,00 €
    1 * 1 ST
       897,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    COMAN2013
    Sensor
    T: Thermistor
    RH: Capacitive
    Display
    LCD
    Battery life
    5 years
    Recording intervals
    1 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
  • COMAN2014
    Temperature data logger, single/multiple thermocouple
    Temperature data logger, single/multiple thermocouple
    T: –200…+1372 °C (K type)
    T: –200…+400 °C (T type)
    T: –40…+70 °C (thermistor)
    T: ±0,5 °C (–20…+40 °C); ±1,0 °C (–40…+70 °C); ±0,5 °C at 20 °C; ±0,3 °C (–20…+40 °C, thermistor); ±0.5 °C (–40…+70 °C, thermistor)
    0,2 °C (0…300 °C); 0,5 °C (>300 °C)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 679,00 €
    679,00 €
    1 * 1 ST
       679,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    COMAN2014
    Sensor
    Int.: Thermistor
    Ext.: Types T/K thermocouple
    Display
    LCD
    Battery life
    5 years
    Recording intervals
    2 s - 99 hr
    Response time
    T90, 15 - 30 min
    W×D×H
    88×80×35 mm
    Data memory - No. of readings
    16000
    Batteries
    1×AA size, 3,6 V lithium battery
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • EV logger software, professional version
    EV logger software, professional version
    COMAEVSWPRO
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Type-K adaptor for data logger, N2014
    Type-K adaptor for data logger, N2014
    COMAN2000ADP/K
    Log in for availability
    1 * 1 ST - 193,00 €
Product Information
  • Catalog #
    Availability
    Price
  • EV logger software, general version
    EV logger software, general version
    COMAEVSW
    Log in for availability
    1 * 1 ST
Product Information
  • Catalog #
    Availability
    Price
  • Type-T adaptor for data logger, N2014
    Type-T adaptor for data logger, N2014
    COMAN2000ADP/T
    Log in for availability
    1 * 1 ST - 206,00 €
Product Information
  • Catalog #
    Availability
    Price
  • USB computer interface
    USB computer interface
    COMAN2000CRU
    Log in for availability
    1 * 1 ST - 114,00 €