Order Entry
Austria
ContactUsLinkComponent
Automated cell counter, EVE™
Automated cell counter, EVE™
Catalog # 734-2675
Supplier:  NANOENTEK
undefined
Automated cell counter, EVE™
Catalog # 734-2675
Supplier:  NANOENTEK

Specifications

  • Model:
    EVE™
  • Description:
    Automatic cell counter incl. power supply and USB drive, 50 slides (for 100 counts) with 1×1,5 ml of trypan blue (0,4%)
  • Seq:
    02
  • Pk:
    1

Specifications

About this item

EVE™ is a benchtop automatic cell counter designed to measure cell count and viability accurately and precisely. It uses the standard trypan blue technique.

  • Fine distinction of clumped cells: Accurate results with advanced analysis algorithm
  • Broad range of cell sizes and types: Primary (tissue and blood) cell lines and stem cells
  • User friendly: Touch screen LCD without any maintenance needed
  • Data store analysis: 500 test results, transfer to PC via USB drive
  • Cell size gating: Select range of cell size using gating function