Order Entry
Austria
ContactUsLinkComponent
VWR®, PCR buffers
VWR®, PCR buffers
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, PCR buffers
Supplier:  VWR Chemicals
Certificates

Specifications

  • Application:
    Standard amplification
  • Storage conditions:
    – 20 °C (long term), + 4 °C (6 months)
  • Concentration:
    10x
  • Description:
    Key Buffer
  • Size:
    3 x 1.5 ml

Specifications

Frequently Bought Together