Orders LinkContactUsLinkComponent
Micro protein quantification set, QuantiMate
Micro protein quantification set, QuantiMate
Supplier:   BIOCHROM
Micro protein quantification set, QuantiMate
Supplier:  BIOCHROM
Certificates
Micro protein quantification set, QuantiMate
Micro protein quantification set, QuantiMate
6 Options Available Below
Product Details & Documents
QuantiMate is designed for concentration and purity determinations of nucleic acids and proteins.
  • Measurement performance giving accurate and reproducible concentration results
  • Samples measured without time consuming dilutions
  • Only one cuvette is required
Dimensions are equivalent to standard cuvettes in order to work in most spectrophotometers.
Product Family Options
Product Information
  • Height
    Volume
    Path length
    Window material
    Availability
    Price
  • 730-1453
    Quantification set
    Quantification set
    8,5 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 730-1454
    QuantiMate 200
    QuantiMate 200
    8,5 mm
    0,8 µl
    200 µm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 730-1455
    QuantiMate 500
    QuantiMate 500
    8,5 mm
    2,5 µl
    500 µm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 730-1450
    Quantification set
    Quantification set
    15 mm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 730-1451
    QuantiMate 200
    QuantiMate 200
    15 mm
    0,8 µl
    200 µm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • 730-1452
    QuantiMate 500
    QuantiMate 500
    15 mm
    2,5 µl
    500 µm
    Quartz
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST