ContactUsLinkComponent
Measuring beakers with handle, PP
Measuring beakers with handle, PP
Catalog # SEMA4732
Supplier:  SEMADENI
Supplier #: 4732
undefined
Measuring beakers with handle, PP
Catalog # SEMA4732
Supplier:  SEMADENI
Supplier #: 4732
Log in for availability
Select an Option
148 mm
Select an Option
20 ml
Select an Option
2000 ml
Select an Option
212 mm
Select an Option
02
Select an Option
1
About this item
PP, highly translucent.
  • Volume is equally visible for left and right handers
  • Good chemical resistance, resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
  • To preserve markings, cleaning at no higher than 60 °C is recommended
  • For autoclaving we recommend the design with raised graduations
Ideal for general laboratory use and a wide variety of other applications. With stable handle, spout and easily readable, printed black scale on both sides. Stackable.
Food-safe product according to EC Directive No. 10/2011
Frequently Bought Together