ContactUsLinkComponent
Wash bottles, wide neck, with vent cap
Wash bottles, wide neck, with vent cap
Catalog # SEMA5264
Supplier:  SEMADENI
undefined
Wash bottles, wide neck, with vent cap
Catalog # SEMA5264
Supplier:  SEMADENI
Specifications
  • Thread:
    GL 63
  • Capacity:
    1000 ml
  • Pk:
    1
  • Imprint:
    Distilled water
Specifications
About this item
Bottles made from LDPE, spray tube from PP.
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No, hazard pictogram, signal word
  • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
With coloured VENT-CAP which supports easy visual identification of the media. Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube. Practically no leakage or dripping with the VENT-CAP, the design of which prevents almost all static overpressure.